missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Clinical Thermometer with Calibration Certificate

Product Code. p-200007085
Click to view available options
Calibrated:
3 Point Traceable
3-point UKAS
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17843018

Brand: Brannan™ 11/064/2 + 49/101/0

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Easy to use waterproof clinical thermometer. Complete with traceable calibration.

  • High accuracy, with fast response
  • Safe, easy to read and simple to use
  • May be used for taking oral, underarm or rectal temperature
  • Temperature range: 32°C to 42.9°C
  • 60 second response time
  • Supplied with calibration
  • Compact, accurate and durable
  • Non-hazardous
  • Complete with battery and instructions

Specifications

Calibration Points 35°C,37°C, 39°C
Certifications/Compliance UK National Standards ISO 9001
For Use With (Application) To take temperature via oral, underarm, or rectal
Length (Metric) 13.3 cm
Material Plastic
Max. Temperature (English) 42.9°F
Max. Temperature (Metric) 42.9°C
Min. Temperature (English) 32°F
Min. Temperature (Metric) 32°C
Temperature Range (English) 32°C to 42.9°C
Temperature Range (Metric) 32°C to 42.9°C
Calibrated 3 Point Traceable
Battery Type 1 x LR41
Accuracy ± 0.1°C between 35.5°C and 42°C, otherwise ± 2°C
Subdivision 0.1°C
Display LCD
Width (Metric) 2.4 cm
IP Rating IP22
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.