Learn More
Thermo Scientific™ ClipTip™ Filtered 384-Format Pipette Tips
Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes.
£779.00 - £794.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Format | Rack |
Packaging | 10 x 384 tips/rack |
Product Code | Brand | Description | Volume (Metric) | Color Code | Fit for Purpose | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Color Code | Fit for Purpose | Price | Quantity & Availability | ||
15942021
|
Thermo Scientific™
94420073 |
ClipTip 384 12.5 Ext, Filtered, Sterile, Rack | 12.5 μL | Pink | Sterile & PCR Ready |
£779.00
Pack of 3840 |
In Stock
Log in to see stock. |
||
15226293
|
Thermo Scientific™
94420053 |
ClipTip 384 12.5, Filtered, Sterile, Rack | 12.5 μL | Pink | Sterile & PCR Ready |
£794.00
Pack of 3840 |
In Stock
Log in to see stock. |
||
15256293
|
Thermo Scientific™
94420103 |
ClipTip 384 30, Filtered, Sterile, Rack | 30 μL | Violet | Sterile & PCR Ready |
£794.00
Pack of 3840 |
In Stock
Log in to see stock. |
||
15286293
|
Thermo Scientific™
94420153 |
ClipTip 384 125, Filtered, Sterile, Rack | 125 μL | Yellow | Sterile & PCR Ready |
£789.00
Pack of 3840 |
In Stock
Log in to see stock. |
||
Description
Thermo Scientific™ ClipTip™ 384-format pipette tips are designed for 384-format microplate applications, in conjunction with the E1-ClipTip electronic pipettes. Featuring an innovative ‘snap and seal’ tip attachment mechanism, the pipette tips ensure a light force with a secure tip attachment, enabling greater efficiency and confidence in your work.
Key Features:
- All ClipTip 384-format tips feature a ‘Snap and Seal’ tip attachment mechanism that ensures light force and secure tip attachment
- Each tip has incorporated small protrusions that snap on the tip fitting flange of the pipette
- Ideal for 384-format microplate applications
- Filtered tips are ideal for sensitive assays, such as PCR, and with critical samples to prevent cross-contaminations
Proven quality
- Certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
- 96-format volumes: 12.5, 20, 50, 200, 300, 1000 and 1250 μL
- 384-format volumes: 12.5, 30 and 125μL
- Extended lenght tips available for volumes 12.5 and 300 μL allowing for better reach with specific labware consumables
Immediate traceability
- Each tip insert has ink-jetted lot and volume information; the convenient color key makes it easy to match the color code of the F1-ClipTip and E1-ClipTip pipette with the corresponding ClipTip pipette tip rack insert
Note:
Availability may vary by country.
Specifications
Filtered | |
Not Autoclavable | |
10 x 384 tips/rack | |
Sterile & PCR Ready |
Sterile | |
Rack | |
ClipTip |