Learn More
Clomazone, Honeywell Fluka™
Brand: Honeywell Fluka R1686-100MG
Additional Details : CAS Number : 81777-89-1 Weight : 0.10000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Clomazone | |
≤ 100% | |
2-[(2-chlorophenyl)methyl]-4,4-dimethyl-3-isoxazolidinone | |
Herbicides | |
32.5°C to 34.2°C | |
Colorless | |
157°C |
81777-89-1 | |
7480026 | |
239.70 g/mol | |
1.192 g/cm3 (20°C) | |
281.7°C (1013 hPa) | |
100 mg |