missing translation for 'onlineSavingsMsg'
Learn More

Clomiphene citrate, MedChemExpress

Product Code. 16420948
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16420948

Brand: Medchem Express HYB0463

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Clomiphene citrate (Clomifene citrate) is a selective estrogen receptor modulator.

Clomiphene citrate (Clomifene citrate) is a selective estrogen receptor modulator.

Specifications

Chemical Name or Material Clomiphene citrate
CAS 50-41-9
Color White
Purity Grade Notes Research
Molecular Formula C32H36ClNO8
Quantity 5 g
Synonym Clomifene citrate
Solubility Information DMSO : ≥ 50 mg/mL (83.60 mM) ∣H2O : 1 mg/mL (1.67 mM; Need ultrasonic)
SMILES Cl/C(C1=CC=CC=C1)=C(C2=CC=CC=C2)/C3=CC=C(OCCN(CC)CC)C=C3.OC(CC(O)=O)(C(O)=O)CC(O)=O
Molecular Weight (g/mol) 598.08
Formula Weight 598.08
Percent Purity 99.69%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.