Learn More
STEMCELL Technologies ClonaCell™-CHO ACF Supplement
Animal component-free, serum-free, glutamine-free 40X supplement for cloning during CHO cell line development
Brand: STEMCELL Technologies 03820
Code : Z1
Additional Details : Weight : 0.00250kg
Description
- ClonaCell™-CHO ACF Supplement is a serum-free and animal component-free supplement that is added to protein-free media in order to increase cloning efficiency and promote robust growth of CHO cells
- The 40X supplement can be added to a variety of protein-free liquid or semi-solid cell culture media to increase the growth and single-cell cloning efficiency of suspension-adapted CHO cells
- The supplement contains recombinant proteins and phenol red, and does not contain animal components, serum, hydrolysates, L-glutamine, or selection agents
Specifications
CHO Cells | |
Cell Culture | |
2.5 mL |