Learn More
STEMCELL Technologies ClonaCell™-CHO CD Liquid Medium
Chemically defined, animal component-free, serum-free, protein-free, glutamine-free liquid medium for culture of CHO cells
Brand: STEMCELL Technologies 03817
Code : Z1
Additional Details : Weight : 0.50000kg
Description
- ClonaCell™-CHO CD Liquid Medium is a general purpose liquid medium for culture of suspension-adapted CHO cells
- This chemically defined liquid medium is protein-free and animal component-free
- It does not contain L-glutamine, selection agents, antibiotics, or phenol red
- ClonaCell™-CHO CD Liquid Medium is compatible with a variety of selection systems, including dihydrofolate reductase (DHFR) and glutamine synthetase (GS)
Specifications
CHO | |
Cell Culture | |
500 mL |