missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CloneMiner™ II cDNA Library Construction Kit
CloneMiner™ II cDNA Library Construction Kit
Brand: Invitrogen™ A11180
2728.40 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.56000kg
Description
The CloneMiner™ II cDNA Library Construction Kit is a second generation CloneMiner™ kit that enables rapid construction of highly representative cDNA libraries without restriction enzyme cloning. This innovative library construction technology combines SuperScript™ III Reverse Transcriptase with Gateway™ cloning technology, resulting in the discovery of previously unobtainable, full-length clones. The kit also avoids the use of time-consuming, inefficient ligation reactions, making library construction faster and offering better representation.The CloneMiner™ II cDNA Library Construction Kit ensures:
How the CloneMiner™ II Kit Works:
How is This Kit Different From the Original CloneMiner™ Kit?
Specifications
Library Preparation Kit | |
DH10B | |
cDNA Library | |
pDONR222 | |
CloneMiner™ | |
1 Kit |
The CloneMiner™ II Kit includes SuperScript™ III RT, Gateway™ vectors, reagents for cDNA library construction and ElectroMax™ DH10B™ T1-Phage-resistant Competent Cells. Please see manual for details. Store ElectroMax™ DH10B™ T1 Phage-Resistant Competent Cells at –80°C, cDNA size fractionation columns at +4°C, all other kit components at –20°C. | |
Gateway | |
SuperScript™ III | |
Kit | |
cDNA Libraries & Library Construction |
For Research Use Only. Not for use in diagnostic procedures.