Learn More
Pro-Lab Diagnostics™ Clostridium Sordelli Sera
Identify the pathogenic Clostridia sordelli or its toxins in pathological material. Pro-Lab Diagnostics™ Clostridium Sordelli Sera is used as a Salmonella agglutinating serum and is used for identification of cultures that have been previously characterized biochemically as Clostridia.
Brand: Pro-Lab Diagnostics™ PL6508
Code : 04
Additional Details : Weight : 1.00000kg
Description
Clostridium sordellii is a rare anaerobic, gram-positive, spore-forming rod with peritrichous flagella that is capable of causing pneumonia, endocarditis, arthritis, peritonitis, and myonecrosis. Clostridia Sera should only be used for identification of cultures which have been previously characterized biochemically as Clostridia. This product is for research use only.Specifications
Salmonella agglutinating serum | |
Identification of Clostridia sordelli or its toxins | |
10 mL | |
Liquid |