Learn More
Thermo Scientific™ Closures with Prefitted Septa for 13mm Crimp/Snap-Ring LoVials™
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
£126.00
Specifications
Closure Size 13 mm
View More Specs
Products 1
Product Code Brand Description Septum Price Quantity & Availability  
Product Code Brand Description Septum Price Quantity & Availability  
11981533
View Documents
Thermo Scientific™
C4015-4A
C4015-4A Red PTFE/White Silicone This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Thermo Scientific™ National Closures with Prefitted Septa are available in easy-to-apply Kim-Snap or standard aluminum variety to suit your application needs.
  • Kim-Snap™ closures provide a tight seal without the need of a crimper
  • National Scientific aluminum crimp closures provide a secure, leak-resistant seal
  • Aluminum seals must be applied with a crimping tool
Specifications
Specifications
13 mm