missing translation for 'onlineSavingsMsg'
Learn More

Clotrimazole, Tocris Bioscience™

Product Code. 18782774
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cytochrome P450 inhibitor; antifungal

Imidazole antimycotic and cytochrome P450 inhibitor. Inhibits Ca2+-activated K+ current (IC50 = 3 μM) and L-type Ca2+ current; increases the firing rate of action potentials. Exhibits antiproliferative activity in vitro and in vivo. Also an inverse agonist of the human constitutive androstane receptor (hCAR).

Specifications

CAS 23593-75-1
Molecular Formula C22H17ClN2
Quantity 50 mg
Synonym clotrimazole, lotrimin, canesten, mycelex, mycosporin, empecid, clotrimazol, mykosporin, gyne lotrimin, chlotrimazole
InChI Key VNFPBHJOKIVQEB-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4
IUPAC Name 1-[(2-chlorophenyl)-diphenylmethyl]imidazole
Molecular Weight (g/mol) 344.842
PubChem CID 2812
ChEBI CHEBI:3764
Formula Weight 344.84
Percent Purity >99%
Chemical Name or Material Clotrimazole
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'