missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Clozapine N-oxide
GREENER_CHOICE

Product Code. 30001355
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001355

Brand: Tocris Bioscience™ 4936/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Activator of muscarinic DREADDs

Metabolite of clozapine (Cat. No. 0444). Activates human muscarinic designer receptors (DREADDs) in vitro. Silences hippocampal neurons expressing hM4 DREADDs in vitro. Activates Gs-linked DREADDS- AgRP in mice via intracerebroventricular (i.c.v) injection. Shown to be a P-glycoprotein (P-gp) efflux pump substrate. Other potent and selective hM3Dq DREADD agonists available: DREADD agonist 21 and Perlapine .

Water soluble salt also available.

This product contains 1 molar equivalent of ethanol.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34233-69-7
Molecular Formula C18H19ClN4O

Specifications

Assay Percent Range >98%
Quantity 10 mg
Formula Weight 342.82
Purity >99%
Molecular Weight (g/mol) 342.827
Chemical Name or Material Clozapine N-oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.