Learn More
Semivolatiles Balance Mix- Option A, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep SV-X
Code : 04
Additional Details : Weight : 0.00100kg Transport : UN number : 1593 Chem class : 6.1 Pack group : III
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
3-Methylphenol | 108-39-4 | 2000μg/mL |
Aniline | 62-53-3 | 2000μg/mL |
Azobenzene | 103-33-3 | 2000μg/mL |
Carbazole | 86-74-8 | 2000μg/mL |
N-Nitrosodimethylamine | 62-75-9 | 2000μg/mL |
Pyridine | 110-86-1 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Semi-Volatile Balance Mix - Option A | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
Aniline; Azobenzene; Carbazole; 3-Methylphenol; N-Nitrosodimethylamine; Pyridine |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
For US EPA Method 8270C | |
MeCl |
Product intended for laboratory use only