Learn More
Volatiles Mix D, High Level, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep CLPV-DH
Code : NEW
Additional Details : CAS Number : 67-56-1 Weight : 0.00100kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
2-Butanone | 78-93-3 | 2000μg/mL |
2-Hexanone | 591-78-6 | 2000μg/mL |
4-Methyl-2-pentanone | 108-10-1 | 2000μg/mL |
Acetone | 67-64-1 | 2000μg/mL |
Carbon disulfide | 75-15-0 | 2000μg/mL |
cis-1,2-Dichloroethene | 156-59-2 | 2000μg/mL |
p-Xylene | 106-42-3 | 2000μg/mL |
Styrene | 100-42-5 | 2000μg/mL |
Vinyl acetate | 108-05-4 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Volatiles Mix D, CLP SOW 2/88 & 3/90 (High Level) | |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
For US EPA CLP Methods | |
Methanol-P&T |
For CLP SOW 2/88 And 3/92 | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
Acetone; 2-Butanone; Carbon Disulfide; cis-1,2-Dichloroethene; 2-Hexanone; 4-Methyl-2-pentanone; Styrene; Vinyl acetate; p-Xylene |
Safety and Handling
- Volatiles Mix D, CLP SOW 2/88 & 3/90 (High Level)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol, hexan-2-one, 4-methylpentan-2-one, carbon disulphide, styrene
Product intended for laboratory use only