Learn More
Invitrogen™ Clusterin Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHCLUX5
Description
The Human Clusterin ELISA quantitates Hu Clusterin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Clusterin. Principle of the method The Human Clusterin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Isoform 1 functions as extracellular chaperone that prevents aggregation of nonnative proteins. Prevents stress-induced aggregation of blood plasma proteins. Inhibits formation of amyloid fibrils by APP, APOC2, B2M, CALCA, CSN3, SNCA and aggregation-prone LYZ variants (in vitro). Does not require ATP. Maintains partially unfolded proteins in a state appropriate for subsequent refolding by other chaperones, such as HSPA8/HSC70. Does not refold proteins by itself. Binding to cell surface receptors triggers internalization of the chaperone-client complex and subsequent lysosomal or proteasomal degradation. Secreted isoform 1 protects cells against apoptosis and against cytolysis by complement. Intracellular isoforms interact with ubiquitin and SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complexes and promote the ubiquitination and subsequent proteasomal degradation of target proteins. Promotes proteasomal degradation of COMMD1 and IKBKB. Modulates NF-kappa-B transcriptional activity. Nuclear isoforms promote apoptosis. Mitochondrial isoforms suppress BAX-dependent release of cytochrome c into the cytoplasm and inhibit apoptosis. Plays a role in the regulation of cell proliferation.Specifications
P10909 | |
15 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
1191 | |
<12% | |
Wash Buffer, SAV-HRP, Stop Solution, Pre-coated 96 well plates, Chromogen, Assay Diluent Concentrate, Standard, Adhesive Plate Covers, Biotinylated Detection Antibody | |
5 x 96 Tests | |
Neurobiology | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
19.53 pg/mL to 20,000 pg/mL | |
15 pg/mL to 20000 pg/mL | |
Biotin | |
Mitochondrial Proteins | |
This ELISA antibody pair detect human Clusterin. Other species not determined. | |
Human | |
Colorimetric Microplate Reader | |
AAG4,APO-J,APOJ,CLI1,CLU2,KUB1,NA1/NA2,SGP-2,SGP2,SP-40,TRPM-2,TRPM2,CLU | |
<10% | |
HRP | |
RUO | |
Plasma, 1 μ;L; Serum, 1 μ;L; Supernatant, 100 μ;L | |
AAG4, APO-J, APOJ, CLI1, CLU2, KUB1, NA1/NA2, SGP-2, SGP2, SP-40, TRPM-2, TRPM2, CLU | |
1 hr. 20 min. |