Learn More
Electrothermal™ CMUA Series Controlled Stirring Mantle
CMUA Electromantles share all the excellent features of the CMU Electromantle, but with the added functionality of magnetic stirring. For convenience and safety, the electric circuitry for stirring is built into the base of the heater, enabling powerful stirring over a wide range of solution volumes and viscosities using a magnetic stir bar.
Brand:  Electrothermal™ 36013-12
 View more versions of this product
Product Code. 12509717
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • The electric circuitry for stirring is built into the base of the heater, enabling powerful stirring over a wide range of solution volumes and viscosities using a magnetic stir bar.
  • Ideal for busy laboratories with multiple users as they are very safe to use
  • Metal case designed to remain cool when in operation.
  • Stir speed is bi-directional and easily adjustable
  • Auto-recapture switch can be engaged in the event a temporarily interruption in magnetic stirring.
Specifications
Specifications
5000 mL
230 V
Powder-coated aluminum
up to 2000rpm
2.75kg
50/60 Hz
320 x 320 x 220mm
230v 50/ 60Hz, 800w, With UK and EU plugs
UK and EU plugs
800W
450°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Electrothermal™ CMUA Series Controlled Stirring Mantle >