missing translation for 'onlineSavingsMsg'
Learn More

Electrothermal™ CMUA Series Controlled Stirring Mantle

CMUA Electromantles share all the excellent features of the CMU Electromantle, but with the added functionality of magnetic stirring. For convenience and safety, the electric circuitry for stirring is built into the base of the heater, enabling powerful stirring over a wide range of solution volumes and viscosities using a magnetic stir bar.

Brand:  Electrothermal™ 36013-06

 View more versions of this product

Product Code. 13137796

  • £920.00 / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The electric circuitry for stirring is built into the base of the heater, enabling powerful stirring over a wide range of solution volumes and viscosities using a magnetic stir bar.
  • Ideal for busy laboratories with multiple users as they are very safe to use
  • Metal case designed to remain cool when in operation.
  • Stir speed is bi-directional and easily adjustable
  • Auto-recapture switch can be engaged in the event a temporarily interruption in magnetic stirring.
Specifications

Specifications

1000 mL
230 V
Powder-coated aluminum
up to 2000rpm
1.75kg
50/60 Hz
220 x 220 x 170mm
230v 50/60Hz, 380w, With UK and EU plugs
UK and EU plugs
380W
450°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers