Learn More
Cytiva CNBr-Activated Sepharose™ 4 Fast Flow Affinity Media
Used for coupling of large amino-containing ligands
Brand: Cytiva 17-0981-01
Code : GE
Additional Details : CAS Number : 9012-36-6 Weight : 0.04320kg
Description
- Resin composed of cross-linked 4% agarose beads that have been pre-activated with cyanogen bromide
- Proteins and other molecules containing primary amino groups are coupled directly to preactivated gel
- Allows for multi-point attachment of protein ligands which minimizes ligand leakage
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
Specifications
10 g | |
Stable in commonly used aqueous Buffers provided that the ligand withstand the pH or the chemical environment | |
CNBr-Activated Sepharose 4 Fast Flow Media | |
45 to 165 μm | |
25 cm | |
100 kPa | |
3 to 11 | |
Cyanogen Bromide Activated |
For Coupling of Large Amino-Containing Ligands | |
2°C to 8°C | |
150 to 250 cm/Hoμr | |
9.84 in. | |
Certificate of Analysis | |
4% Cross-Linked Agarose | |
20% Ethanol |