Learn More
CNX-2006, MedChemExpress
MedChemExpress CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.
Brand: Medchem Express HY-13897
This item is not returnable.
View return policy
Description
CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.Specifications
CNX-2006 | |
Gray | |
C26H27F4N7O2 | |
DMSO : ≥ 52 mg/mL (95.32 mM) | |
545.53 | |
99.68% | |
Solid |
1375465-09-0 | |
Research | |
5 mg | |
C=CC(NC1=CC=CC(NC2=NC(NC3=CC=C(NC4CN(CCF)C4)C=C3OC)=NC=C2C(F)(F)F)=C1)=O | |
545.53 | |
Research | |
Cancer-Kinase/protease |