missing translation for 'onlineSavingsMsg'
Learn More

Co 101244 hydrochloride, Tocris Bioscience™

Product Code. 18034592
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GluN2B-selective NMDA antagonist

Novel, potent and selective antagonist of GluN2B (formally NR2B)-containing NMDA receptors (IC50 values are 0.043, > 100 and > 100 μM for GluN1A/2B (NR1A/2B), GluN1A/2A (NR1A/2A) and GluN1A/2C (NR1A/2C) subunit combinations respectively). Displays neuroprotective effects in vivo and in vitro.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 193356-17-1
Molecular Formula C21H28ClNO3
Molecular Weight (g/mol) 377.909
InChI Key WFFZHKKSIDENAJ-UHFFFAOYSA-N
Synonym co 101244 hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl piperidin-4-ol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol mono hydrochloride, d08nrh, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol monohydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-4-methylphenyl methyl-4-piperidinol hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4-hydroxy-4-4-methylbenzyl piperidine hydrochloride, 1-2-4-hydroxyphenoxy ethyl-4hydroxy-4-4-methylbenzyl piperidine hydrochloride
PubChem CID 6918426
IUPAC Name 1-[2-(4-hydroxyphenoxy)ethyl]-4-[(4-methylphenyl)methyl]piperidin-4-ol;hydrochloride
SMILES CC1=CC=C(C=C1)CC2(CCN(CC2)CCOC3=CC=C(C=C3)O)O.Cl

Specifications

Quantity 50 mg
Formula Weight 377.91
Percent Purity >99%
Chemical Name or Material Co 101244 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'