Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Brand:  Thermo Scientific™ CO4567
1987.70 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.
 View more versions of this product
Product Code. 11334222
  • £2144.41 / Each
Estimated Shipment: 07-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:
  • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
  • CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CO2 Backup, Factory Installed
-80°C
TSX Series, Forma FDE, HERAfreeze HDE, Revco RDE Series, Revco TDE Series
Boxes (300)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers > CO2 Backup System; Factory Installed; For use with Forma 88000, HERAfreeze HFU-T, Revco UxF and TSU Series