Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Brand:  Thermo Scientific™ 1960446
Additional Details : Weight : 0.99000kg
 View more versions of this product
Product Code. 16349693
  • £2004.94 / Each
Estimated Shipment: 23-10-2024
to see stock.
Add to basket
Description
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:
  • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
  • CO2 backup system maintains temperature down to -7° or -70°C (model-dependent) with CO2
Ordering Information:
Available as field- or factory-installed. Models for UxF300/88300/HFU300T/TSU300 Series are standalone. Field-installed units must be installed by qualified professionals.
Specifications
Specifications
CO2 Backup System
Field-Installed
-80°C
TSE Series, Forma™ 7000 Series, HFU B and HFC Series, Revco™ CxF Series, TSC Series