Learn More
Fisherbrand™ CO2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp CO2 Backup Systems for Ultra-Low Temperature Freezers.
Brand: Fisherbrand™
Additional Details : Weight : 0.10000kg
Description
- Maintains temperature down to -67°C with CO2
Specifications
Factory Installed | |
-67°C | |
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer |