missing translation for 'onlineSavingsMsg'
Learn More
Carbon Dioxide Standard, 2g/L, Reagecon™ Product Code.: 11708465

Carbon Dioxide Standard, 2g/L, Reagecon™
BIOPHARMA

Product Code. 11708465 Shop All Reagecon Products
100 Ampules
Click to view available options
Quantity:
100 Ampules
This item is not returnable. View return policy

Product Code. 11708465

Brand: Reagecon SC022C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Packaged in individual 2mL ampules. Carbon Dioxide Standard, 2g/L, Reagecon™ is a 2g/L CO2 release agent used for calibration and control purposes with the M965D Carbon Dioxide Analyzer and in soft drink and brewery applications. Traceable to NIST.

Solution of 2g/L aqueous carbon dioxide packaged in individual 2mL sealed glass vials.

Specifications

CAS 124-38-9, 7732-18-5
Packaging 2mL glass ampules
Quantity 100 Ampules
Vapor Pressure 23hPa at 20°C
Density 1.00739g/cm3
Molecular Formula CO2
Physical Form Liquid
Concentration 2 g/L
Synonym Carbonic acid
Color Colorless
InChI Key CURLTUGMZLYLDI-UHFFFAOYSA-N
Melting Point 0°C
Boiling Point 100°C
SMILES C(=O)=O
IUPAC Name carbon dioxide
Solubility Miscible with water
Molecular Weight (g/mol) 44.009
PubChem CID 280
ChEBI CHEBI:16526
Formula Weight 44.01g/mol
Chemical Name or Material Carbon Dioxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC02223G1 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberSC022C
Lot NumberC02222G1 Certificate TypeCertificate of Analysis Date28/02/2023 Catalog NumberSC022C
Lot NumberC02220H1 Certificate TypeCertificate of Analysis Date01/07/2021 Catalog NumberSC022C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carbon Dioxide Standard, 2g/L, Reagecon™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.