missing translation for 'onlineSavingsMsg'
Learn More

Biochrom CO7500 Filter 650 nm

CO7500 Filter 650 nm is an additional wavelength filter for users with Libra S2 and WPA CO7500 colorimeters.

Brand:  Biochrom 80-3003-18

Additional Details : Weight : 0.10000kg

Product Code. 17460663

  • £194.00 / Each
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

  • Libra S2 and WPA CO7500 colorimeters
Specifications

Specifications

650 nm
40 nm
Biochrom CO7000, CO7500 colorimeters
Spectro
Colorimeter Filter
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers