missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coagulation Factor III/ Tissue Factor Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 5 publications
£163.06 - £469.23
Specifications
Antigen | Coagulation Factor III/Tissue Factor |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728157
|
R&D Systems
AF2339-SP |
25 μg |
£163.06
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18001952
|
R&D Systems
AF2339 |
100 μg |
£469.23
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coagulation Factor III/Tissue Factor Polyclonal specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.Specifications
Coagulation Factor III/Tissue Factor | |
Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
F3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF | |
Unconjugated | |
RUO | |
P13726 | |
2152 | |
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III/Tissue Factor Gly34-Glu251 Accession # P13726 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Coagulation Factor III/Tissue Factor in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse Coagulation Factor III/Tissue Factor is observed. |
For Research Use Only