Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters.
Brand:  Buerkle™ 0600-0120
 View more versions of this product
Product Code. 10030042
  • £16.85 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • All adapters have a standard outer thread R 3/4 in.
Specifications
Specifications
Thread Adapter
Polypropylene, Polyethylene
3/4 in. Inflowing, 2 in. Mauser outflowing
Green/White
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in. > Thread Size: 3/4 in. Inflowing, 2 in. Mauser outflowing