Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters.
Brand: Buerkle™ 0600-0090
6.51 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- All adapters have a standard outer thread R 3/4 in.
Specifications
Thread Adapter | |
PVC | |
3/4 in. Captive Cap | |
Grey |
Product Content Correction
Product Title
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in. > Captive cap for 3/4 in. Outflowing
Spot an opportunity for improvement?Share a Content Correction