Learn More
BINDER™ Coated Heat Exchanger
Heat exchanger that is coated in plastic to prevent damage.
Brand:  BINDER™ 8012-2463
 View more versions of this product
Product Code. 30238248
  • £864.00 / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
Specifications
KB PRO 470
Aluminum
Heat Exchanger
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Coated Heat Exchanger > For use with KB PRO 470