Learn More
BINDER™ Coated Heat Exchanger
Product Code. 30238380 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB PRO 1600
KB PRO 260
KB PRO 470
KB PRO 720, KB PRO 1060
KBF PRO 130
KBF PRO 1600
KBF PRO 260
KBF PRO 470
KBF PRO 720, KBF PRO 1060
Unit Size:
Each
Product Code. 30238380
Brand: BINDER™ 80122581
Only null left
Add to basket
Add to basket
Heat exchanger that is coated in plastic to prevent damage.
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
For Use With (Equipment) KB PRO 1600
Material Aluminum
Type Heat Exchanger