missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments Coated Receiving Flask

For use with rotary evaporators

£108.00 - £262.00

Specifications

Color Clear
Material Glass
Type Receiving Flask
For Use With (Equipment) Rotary evaporators
Material (Coating) Transparent plastic
View More Specs

Products 5
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10351442
View Documents
Heidolph Instruments
514-81000-02
100 mL
£108.00
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
10147701
View Documents
Heidolph Instruments
514-82000-02
250 mL
£115.00
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
 
10438781
View Documents
Heidolph Instruments
514-83000-02
500 mL
£129.00
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
 
10381532
View Documents
Heidolph Instruments
514-84000-02
1000 mL
£166.00
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
10779562
View Documents
Heidolph Instruments
514-87000-02
3000 mL
£262.00
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
 
Description

Description

  • With S 35/20
  • With transparent plastic coating
Specifications

Specifications

Clear
Receiving Flask
Transparent plastic
Glass
Rotary evaporators
35/20
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heidolph Instruments Coated Receiving Flask

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.