Learn More
Thermo Scientific™ ELISA Carbonate Coating Buffer
For use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA
Brand: Thermo Scientific™ CB01100
57.48 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
- ELISA Carbonate Coating Buffer is intended for use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA.
- The buffer is a 50 mM carbonate buffer, pH 9.4.

Specifications
Coating Buffer | |
Enzyme Labeled | |
ELISA |
100 mL | |
Concentrate | |
Buffer. Store at 2°C to 8°C. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
440160-000 | Certificate of Analysis | 13/03/2025 | |
440161-000 | Certificate of Analysis | 13/03/2025 | |
437797-000 | Certificate of Analysis | 13/03/2025 | |
437428-000 | Certificate of Analysis | 10/03/2025 | |
437428-000 | Certificate of Analysis | 25/01/2025 | CB01100 |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Your input is important to us. Please complete this form to provide feedback related to the content on this product.