missing translation for 'onlineSavingsMsg'
Learn More

Cobalt, 99+%, Thermo Scientific Chemicals

Product Code. 15229846
1 kg
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Melting Point 1495.0°C
Boiling Point 2870.0°C
Quantity 1 kg
Solubility Information Solubility in water: insoluble.
Formula Weight 58.93
Percent Purity ≥99%
Grade Pure
Chemical Name or Material Cobalt
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt, powder
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.