missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt foil, 0.5mm (0.02in) thick, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 x 50 mm
50 x 100 mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 25 x 50 mm |
Percent Purity | 99.995% |
Grade | Puratronic™ |
Odor | Odorless |
Weight | ≈5.6g/25x50mm |
Form | Foil |
Assay | (metals basis) |
Chemical Name or Material | Cobalt foil, 0.5mm (0.02 in.) thick |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X08K018 | Certificate of Analysis | 16/12/2024 | 011451.FG,011451.FY |
R14K018 | Certificate of Analysis | 11/06/2024 | 011451.FG,011451.FY |
X11I023 | Certificate of Analysis | 08/04/2024 | 011451.FG,011451.FY |
X19G063 | Certificate of Analysis | 23/12/2022 | 011451.FG,011451.FY |
P29E060 | Certificate of Analysis | 08/12/2021 | 011451.FG,011451.FY |
Safety and Handling


Product Identifier
- Cobalt foil, 0.5mm (0.02in) thick, Puratronic
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1B
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Cobalt
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cobalt foil, 0.5mm (0.02in) thick, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction