missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Quantity | 25 x 25 mm |
| Odor | Odorless |
| Weight | ≈5.6g/25x25mm |
| Form | Foil |
| Percent Purity | 99.95% |
| Assay | (metals basis) |
| Chemical Name or Material | Cobalt foil, 1.0mm (0.04 in.) thick |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido