Learn More
Cobalt ICP Standard, Fluka™
Certified Reference Material, 10,000 mg/L Co in HNO3 5%
Brand: Honeywell Fluka E1141-100ML
Additional Details : CAS Number : 7697-37-2 Weight : 0.10000kg
Description
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Cobalt ICP Standard | |
100 mL | |
<2 |
Plastic Bottle | |
ICP | |
ICP-OES and ICP-MS |