missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) 2,4-pentanedionate, Thermo Scientific Chemicals

Product Code. 11377377
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cobalt(II) 2,4-pentanedionate is used as an important catalyst for organic reactions. It is also used as paint dryer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-48-7
Molecular Formula C10H14CoO4
Molecular Weight (g/mol) 257.15
MDL Number MFCD00000014
InChI Key UTYYEGLZLFAFDI-FDGPNNRMSA-L
Synonym Cobalt(II) acetylacetonate; Bis(acetylacetonato)cobalt(II)
IUPAC Name λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 165°C to 170°C
Color Blue-Purple
Odor Odorless
Quantity 250 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 257.14
Physical Form Crystalline
Chemical Name or Material Cobalt(II) 2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.