Learn More
Cobalt(II) 2,4-pentanedionate, Thermo Scientific Chemicals
£50.50 - £156.00
Chemical Identifiers
CAS | 14024-48-7 |
---|---|
Molecular Formula | C10H14CoO4 |
Molecular Weight (g/mol) | 257.15 |
MDL Number | MFCD00000014 |
InChI Key | UTYYEGLZLFAFDI-FDGPNNRMSA-L |
Synonym | Cobalt(II) acetylacetonate; Bis(acetylacetonato)cobalt(II) |
IUPAC Name | λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate) |
SMILES | [Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367377
|
Thermo Scientific Alfa Aesar
012537.18 |
50 g |
£50.50
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377377
|
Thermo Scientific Alfa Aesar
012537.30 |
250 g |
£156.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) 2,4-pentanedionate is used as an important catalyst for organic reactions. It is also used as paint dryer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14024-48-7 | |
257.15 | |
UTYYEGLZLFAFDI-FDGPNNRMSA-L | |
λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate) |
C10H14CoO4 | |
MFCD00000014 | |
Cobalt(II) acetylacetonate; Bis(acetylacetonato)cobalt(II) | |
[Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Specifications
14024-48-7 | |
Blue-Purple | |
C10H14CoO4 | |
Hygroscopic | |
Soluble in water. | |
[Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
257.15 | |
Crystalline |
165°C to 170°C | |
Odorless | |
MFCD00000014 | |
Cobalt(II) acetylacetonate; Bis(acetylacetonato)cobalt(II) | |
UTYYEGLZLFAFDI-FDGPNNRMSA-L | |
λ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate) | |
257.14 | |
Cobalt(II) 2,4-pentanedionate |
Safety and Handling
GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P501c
H302-H317-H318-H334-H341-H350
missing translation for 'einecsNumber' : 237-855-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Cobalt(II) 2,4-pentanedionate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Cobalt(II) 2,4-pentanedionate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.