missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate tetrahydrate, 99.999% (metals basis), Thermo Scientific Chemicals

£59.19 - £905.51

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
Molecular Weight (g/mol) 249.081
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
IUPAC Name cobalt(2+);diacetate;tetrahydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11418480
View Documents
Thermo Scientific Alfa Aesar
044345.04
2 g
£59.19
2g
Please to purchase this item. Need a web account? Register with us today!
11408480
View Documents
Thermo Scientific Alfa Aesar
044345.09
10 g
£229.14
10g
Please to purchase this item. Need a web account? Register with us today!
11498470
View Documents
Thermo Scientific Alfa Aesar
044345.18
50 g
£905.51
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cobalt(II) acetate tetrahydrate is used to prepare complexes for examination of the properties of metals with unusual coordination geometries. It is also used as a catalyst for oxidation and esterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6147-53-1
249.081
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
61372
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
C4H14CoO8
MFCD00149569
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
cobalt(2+);diacetate;tetrahydrate

Specifications

∼140°C (in -4H2O)
Red
C4H14CoO8
MFCD00149569
3731397
cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
ZBYYWKJVSFHYJL-UHFFFAOYSA-L
cobalt(2+);diacetate;tetrahydrate
61372
99.999%
1.542
1.71 g/mL
Cobalt (II) acetate tetrahydrate
6147-53-1
Crystalline
Co(OOCCH3)2·4H2O
UN3077
14,2433
Soluble in water(380g/L).
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
249.081
249.08 (177.02 Anhydrous)
Odorless
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

P201-P202-P261-P271-P272-P280g-P281-P285-P302+P352-P304+P340-P308+P313-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H335-H341-H350i-H360F

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 200-755-8

missing translation for 'rtecsNumber' : AG3325000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only