missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) acetate tetrahydrate, Co 24%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
It is used as an industrial catalyst that allow paints and varnishes to harden. Also as a precursor to various oil drying agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | ∽140°C-4H2O |
Color | Red |
Physical Form | Crystals |
Quantity | 250 g |
Assay Percent Range | 24% |
Beilstein | 3731397 |
Solubility Information | Soluble in water,alcohol,dilute acids |
Formula Weight | 249.08 (177.02 Anhydrous) |
Odor | Odorless |
Refractive Index | 1.542 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'