missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) bromide, ultra dry, 99.99% (metals basis)

Product Code. 11303888
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11303888

Brand: Thermo Scientific Alfa Aesar 040319.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-43-7 | Br2Co | 218.741 g/mol

Used primarily as a catalyst in some processes. Chiefly used in hygrometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-43-7
Molecular Formula Br2Co
Molecular Weight (g/mol) 218.741
MDL Number MFCD00016017
InChI Key BZRRQSJJPUGBAA-UHFFFAOYSA-L
Synonym cobalt bromide, cobalt ii bromide, cobalt bromide cobr2, cobalt dibromide, cobaltous bromide, hsdb 998, cobalt bromide ous
PubChem CID 24610
IUPAC Name dibromocobalt
SMILES [Co](Br)Br

Specifications

Melting Point 678°C (in N2)
Color Green
Physical Form Beads
Quantity 5 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water. Soluble in methyl acetate,ether,alcohol,acetone.
Formula Weight 218.75
Sensitivity Hygroscopic
Appearance Green (powder/flake/crystalline/beads, etc.)
Density 4.909
Chemical Name or Material Cobalt(II) bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.