missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) carbonate, 99.5% (metals basis), Thermo Scientific Chemicals

£61.00 - £191.96

Chemical Identifiers

CAS 513-79-1
Molecular Formula CCoO3
Molecular Weight (g/mol) 118.94
MDL Number MFCD00010940
InChI Key ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
Synonym cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuvShow MoreShow Less
PubChem CID 10565
IUPAC Name cobalt(2+);carbonate
SMILES [Co++].[O-]C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11335547
SDS View Documents
Thermo Scientific Alfa Aesar
042745.18
50 g
£61.00
50g
Please to purchase this item. Need a web account? Register with us today!
11325547
SDS View Documents
Thermo Scientific Alfa Aesar
042745.30
250 g
£191.96
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cobalt(II) carbonate is a precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and is also used in the manufacture of paints and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c

H302-H317-H334-H341-H350i-H360F

missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 208-169-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) carbonate, 99.5% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.