missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II) carbonate, 99% (metals basis), Co 45% min, Thermo Scientific Chemicals Product Code.: 11391109

Cobalt(II) carbonate, 99% (metals basis), Co 45% min, Thermo Scientific Chemicals

Product Code. 11391109
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11391109

Brand: Thermo Scientific Alfa Aesar 011343.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use in Ceramics, Manufacturing Co Compounds, Pigments

Cobalt(II) carbonate is precursor to cobalt carbonyl, cobalt salts and blue pottery glazes such as delftware. It is used as an essential element in dietary supplements and also used in the manufacture of paints and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-79-1
Molecular Formula CCoO3
Molecular Weight (g/mol) 118.94
MDL Number MFCD00010940
InChI Key ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
Synonym cobalt carbonate, cobalt ii carbonate, cobaltous carbonate, sphaerocobaltite, cobalt spar, cobalt 2+ carbonate, cobalt monocarbonate, cobalt carbonate 1:1, cobalt carbonate coco3, unii-7h73a68fuvShow MoreShow Less
PubChem CID 10565
IUPAC Name cobalt(2+);carbonate
SMILES [Co++].[O-]C([O-])=O

Specifications

Melting Point (decomposition)
Color Pink
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% (Metals basis, Co 45% min)
Solubility Information Insoluble in water,alcohol and methyl acetate.
Formula Weight 118.94
Odor Odorless
Density 4.13
Chemical Name or Material Cobalt(II) carbonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ23I006 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number011343.36,011343.22
Lot NumberZ23I005 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number011343.36,011343.22
Lot NumberY03A020 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011343.36,011343.22
Lot NumberE10E03 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011343.36,011343.22
Lot NumberJ15N11 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011343.36,011343.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) carbonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Carbonic acid, cobalt(2+) salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) carbonate, 99% (metals basis), Co 45% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.