Learn More
Cobalt(II) chloride, anhydrous, 97%, Thermo Scientific Chemicals
£24.82 - £174.87
Chemical Identifiers
CAS | 7646-79-9 |
---|---|
Molecular Formula | Cl2Co |
Molecular Weight (g/mol) | 129.83 |
MDL Number | MFCD00010938 |
InChI Key | GVPFVAHMJGGAJG-UHFFFAOYSA-L |
Synonym | cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride |
IUPAC Name | λ²-cobalt(2+) dichloride |
SMILES | [Cl-].[Cl-].[Co++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439517
|
Thermo Scientific Alfa Aesar
B22031.14 |
25 g |
£24.82
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449517
|
Thermo Scientific Alfa Aesar
B22031.22 |
100 g |
£57.49
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459517
|
Thermo Scientific Alfa Aesar
B22031.36 |
500 g |
£174.87
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.
Solubility
Freely soluble in water and methanol. Soluble in alcohol, acetone, ether, glycerol, and pyridine.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
7646-79-9 | |
129.83 | |
GVPFVAHMJGGAJG-UHFFFAOYSA-L | |
λ²-cobalt(2+) dichloride |
Cl2Co | |
MFCD00010938 | |
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride | |
[Cl-].[Cl-].[Co++] |
Specifications
1049°C | |
7646-79-9 | |
CoCl2 | |
UN3077 | |
cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride | |
GVPFVAHMJGGAJG-UHFFFAOYSA-L | |
λ²-cobalt(2+) dichloride | |
129.84 | |
Odorless | |
3.367 |
735°C | |
Cl2Co | |
MFCD00010938 | |
14,2437 | |
Freely soluble in water and methanol. Soluble in alcohol,acetone,ether,glycerol,and pyridine. | |
[Cl-].[Cl-].[Co++] | |
129.83 | |
97% | |
Hygroscopic | |
Cobalt(II) chloride, anhydrous |
Safety and Handling
GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P501c
H302+H332-H317-H318-H334-H335-H341-H350i-H360F-H373
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 231-589-4
missing translation for 'rtecsNumber' : GF9800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Product Description: Cobalt(II) chloride, anhydrous
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Cobalt chloride (CoCl2)
RUO – Research Use Only