missing translation for 'onlineSavingsMsg'
Learn More
Cobalt(II) chloride hexahydrate, 98%, Thermo Scientific Chemicals Product Code.: 11437817

Cobalt(II) chloride hexahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11437817
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11437817

Brand: Thermo Scientific Alfa Aesar A16346.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cobalt(II) chloride hexahydrate is widely used as a drying agent in painting and coloring agents for glass ceramics. It is an excellent indicator for water in dessicants as well as an indicator for silica gel dryer. It helps in electroplating. It acts as an inducer of HIF-1 production used to study apoptotic effects in HepG2 cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) chloride hexahydrate is widely used as a drying agent in painting and coloring agents for glass ceramics. It is an excellent indicator for water in dessicants as well as an indicator for silica gel dryer. It helps in electroplating. It acts as an inducer of HIF-1 production used to study apoptotic effects in HepG2 cells.

Solubility
Soluble in water, ether, acetone and alcohol.

Notes
Incompatible with strong oxidizing agents and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobaltous chloride hexahydrate
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 87°C
pH 4.9
Quantity 50 g
Linear Formula CoCl2·6H2O
UN Number UN3077
Merck Index 14,2437
Solubility Information Soluble in water,ether,acetone and alcohol.
Formula Weight 237.93 (129.84 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 1.924
Chemical Name or Material Cobalt(II) chloride hexahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10254782 Certificate TypeCertificate of Analysis Date11/04/2025 Catalog NumberA16346.18,A16346.0B,A16346.30
Lot Number10251905 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog NumberA16346.18,A16346.0B,A16346.30
Lot Number10250160 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog NumberA16346.18,A16346.0B,A16346.30
Lot Number10249319 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog NumberA16346.18,A16346.0B,A16346.30
Lot Number10247346 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog NumberA16346.18,A16346.0B,A16346,A16346.30
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) chloride hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) chloride hexahydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.