missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals
Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 86.0°C |
Color | Purple to Red |
Physical Form | Powder |
Quantity | 100 g |
Linear Formula | CoCl2·6H2O |
Fieser | 01,155; 06,127; 11,479 |
Merck Index | 15, 2421 |
Solubility Information | Solubility in water: 970g/L (20°C) |
Formula Weight | 237.93 |
Percent Purity | 98.0 to 102.0% (Complexometry) |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464084 | Certificate of Analysis | 04/07/2024 | 192092500,192090010,192091000 |
A0458386 | Certificate of Analysis | 04/12/2023 | 192092500,192090010,192091000 |
A0452304 | Certificate of Analysis | 02/05/2023 | 192092500,192090010,192091000 |
A0383284 | Certificate of Analysis | 12/12/2022 | 192092500,192090010,192091000 |
A0418048 | Certificate of Analysis | 12/12/2022 | 192092500,192090010,192091000 |
Safety and Handling



Product Identifier
- Cobalt(II) chloride hexahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H341-Suspected of causing genetic defects.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cobalt(II) chloride hexahydrate, for analysis, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction