missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Chloride Hexahydrate, Certified AR for Analysis, Fisher Chemical™

£245.00 - £4195.00

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10716671
SDS missing translation for 'viewDocuments'
Fisher Chemical
C/6560/48
100 g
£245.00
100g
Please to purchase this item. Need a web account? Register with us today!
10273610
SDS missing translation for 'viewDocuments'
Fisher Chemical
C/6560/53
500 g
£614.64
500g
Please to purchase this item. Need a web account? Register with us today!
10375550
SDS missing translation for 'viewDocuments'
Fisher Chemical
C/6560/60
1 kg
£1175.00
1kg
Please to purchase this item. Need a web account? Register with us today!
12668625
SDS missing translation for 'viewDocuments'
Fisher Chemical
C/6560/65
5 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

7791-13-1
237.92
GFHNAMRJFCEERV-UHFFFAOYSA-L
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Cl2CoH12O6
MFCD00149652
λ²-cobalt(2+) hexahydrate dichloride

Specifications

86°C
Red to Violet
Crystalline Solid
MFCD00149652
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
237.92
Plastic powder jar
7791-13-1
4.6
Cl2CoH12O6
GFHNAMRJFCEERV-UHFFFAOYSA-L
λ²-cobalt(2+) hexahydrate dichloride
237.93g/mol
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt(II) Chloride Hexahydrate, Certified AR for Analysis, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.