Learn More
Cobalt(II) chloride hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Cobalt(II) chloride hexahydrate has been used to study apoptotic effects in HepG2 cells. Also reported as a stabilizer for reconstitution of the radiopharmaceutical technetium-99m exametazime. It has been used in studies to examine heat shock protein expression of inflamed skin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 87°C |
Color | Purple-Red |
pH | 4.9 |
Physical Form | Crystalline Aggregates |
Quantity | 10 g |
Linear Formula | CoCl2·6H2O |
UN Number | UN3077 |
Merck Index | 14,2437 |
Solubility Information | Soluble in water,acetone,ether,and alcohols. |
Formula Weight | 237.93 (129.84 Anhydrous) |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S06K005 | Certificate of Analysis | 12/06/2024 | 010692.18,010692.09 |
M08F068 | Certificate of Analysis | 11/11/2022 | 010692.18,010692.09 |
X19I049 | Certificate of Analysis | 26/10/2022 | 010692.18,010692.09 |
Z21H036 | Certificate of Analysis | 12/04/2022 | 010692.18,010692.09 |
H01Y022 | Certificate of Analysis | 12/11/2021 | 010692.18,010692.09 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.