Learn More
Cobalt(II) chloride hexahydrate, pure, Thermo Scientific Chemicals
£60.60 - £205.00
Chemical Identifiers
CAS | 7791-13-1 |
---|---|
Molecular Formula | Cl2CoH12O6 |
Molecular Weight (g/mol) | 237.92 |
MDL Number | MFCD00149652 |
InChI Key | GFHNAMRJFCEERV-UHFFFAOYSA-L |
IUPAC Name | λ²-cobalt(2+) hexahydrate dichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15269666
|
Thermo Scientific Acros
192862500 |
250 g |
£60.60
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15259666
|
Thermo Scientific Acros
192860010 |
1 kg |
£205.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7791-13-1 | |
237.92 | |
GFHNAMRJFCEERV-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Cl-].[Cl-].[Co++] |
Cl2CoH12O6 | |
MFCD00149652 | |
λ²-cobalt(2+) hexahydrate dichloride |
Specifications
86.0°C | |
Cl2CoH12O6 | |
MFCD00149652 | |
15,2421 | |
GFHNAMRJFCEERV-UHFFFAOYSA-L | |
λ²-cobalt(2+) hexahydrate dichloride | |
237.93 | |
Pure |
7791-13-1 | |
CoCl2·6H2O | |
01,155; 06,127; 11,479 | |
Solubility in water: 970g/L (20°C). | |
O.O.O.O.O.O.[Cl-].[Cl-].[Co++] | |
237.92 | |
98.0 to 102.0% (Complexometry) | |
Cobalt(II) chloride hexahydrate |
Safety and Handling
GHS H Statement
May damage fertility.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing genetic defects.
May cause cancer by inhalation.
May cause an allergic skin reaction.
May cause allergy or ast
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
GHS Signal Word: Danger
- Cobalt(II) chloride hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H341-Suspected of causing genetic defects.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only