missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride, Honeywell™

Product Code. 15666000
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a., anhydrous,≥98.0% (KT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-79-9
Molecular Formula Cl2Co
Molecular Weight (g/mol) 129.83
MDL Number MFCD00010938
InChI Key GVPFVAHMJGGAJG-UHFFFAOYSA-L
Synonym cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride
IUPAC Name λ²-cobalt(2+) dichloride
SMILES [Cl-].[Cl-].[Co++]

Specifications

Quantity 250 g
Linear Formula CoCl2
UN Number UN3077
Formula Weight 129.84g/mol
Percent Purity ≥98% (KT)
Vapor Pressure 40 mmHg (0°C)
Chemical Name or Material Cobalt(II) chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.