Learn More
Cobalt(II) chromate, Thermo Scientific™
£47.00 - £133.00
Chemical Identifiers
CAS | 13455-25-9 |
---|---|
Molecular Formula | CoCrO4 |
Molecular Weight (g/mol) | 174.93 |
MDL Number | MFCD00053962 |
InChI Key | XTUHPOUJWWTMNC-UHFFFAOYSA-N |
Synonym | cobalt ii chromate, cobalt 2+ ; dioxido dioxo chromium, cobaltous chromate iii, cobalt chromate ous, cobaltous chromate, acmc-1c0r0, cobalt 2+ chromate 2-, chromic acid h2cro4 , cobalt 2+ salt 1:1Show More |
PubChem CID | 61614 |
IUPAC Name | cobalt(2+);dioxido(dioxo)chromium |
SMILES | [Co++].[O-][Cr]([O-])(=O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
13455-25-9 | |
174.93 | |
XTUHPOUJWWTMNC-UHFFFAOYSA-N | |
61614 | |
[Co++].[O-][Cr]([O-])(=O)=O |
CoCrO4 | |
MFCD00053962 | |
cobalt ii chromate, cobalt 2+ ; dioxido dioxo chromium, cobaltous chromate iii, cobalt chromate ous, cobaltous chromate, acmc-1c0r0, cobalt 2+ chromate 2-, chromic acid h2cro4 , cobalt 2+ salt 1:1Show More | |
cobalt(2+);dioxido(dioxo)chromium |
Specifications
(decomposition) | |
Lump | |
MFCD00053962 | |
cobalt ii chromate, cobalt 2+ ; dioxido dioxo chromium, cobaltous chromate iii, cobalt chromate ous, cobaltous chromate, acmc-1c0r0, cobalt 2+ chromate 2-, chromic acid h2cro4 , cobalt 2+ salt 1:1 | |
XTUHPOUJWWTMNC-UHFFFAOYSA-N | |
cobalt(2+);dioxido(dioxo)chromium | |
61614 | |
Odorless |
13455-25-9 | |
CoCrO4 | |
UN3087 | |
Soluble in alkali and ammomium hydroxide | |
[Co++].[O-][Cr]([O-])(=O)=O | |
174.93 | |
174.93 | |
Cobalt(II) chromate |
Safety and Handling
GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P210-P220-P221-P260-P264b-P271-P272-P280g-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P370+P378q-P501c
H272-H314-H317-H334-H335-H350i
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II
missing translation for 'einecsNumber' : 236-651-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.